Merck
  • Evidence for distinct pathomechanisms in B-cell chronic lymphocytic leukemia and mantle cell lymphoma by quantitative expression analysis of cell cycle and apoptosis-associated genes.
Evidence for distinct pathomechanisms in B-cell chronic lymphocytic leukemia and mantle cell lymphoma by quantitative expression analysis of cell cycle and apoptosis-associated genes.
Blood (2002-05-31)
Christian Korz, Armin Pscherer, Axel Benner, Daniel Mertens, Claudia Schaffner, Elke Leupolt, Hartmut Döhner, Stephan Stilgenbauer, Peter Lichter
ZUSAMMENFASSUNG
The B-cell lymphoproliferative malignancies B-cell chronic lymphocytic leukemia (B-CLL) and mantle cell lymphoma (MCL) share characteristics, including overlapping chromosomal aberrations with deletions on chromosome bands 13q14, 11q23, 17p13, and 6q21 and gains on chromosome bands 3q26, 12q13, and 8q24. To elucidate the biochemical processes involved in the pathogenesis of B-CLL and MCL, we analyzed the expression level of a set of genes that play central roles in apoptotic or cell proliferation pathways and of candidate genes from frequently altered genomic regions, namely ATM, BAX, BCL2, CCND1, CCND3, CDK2, CDK4, CDKN1A, CDKN1B, E2F1, ETV5, MYC, RB1, SELL, TFDP2, TNFSF10, and TP53. Performing real-time quantitative reverse transcription polymerase chain reaction in a panel of patients with MCL and B-CLL and control samples, significant overexpression and underexpression was observed for most of these genes. Statistical analysis of the expression data revealed the combination of CCND1 and CDK4 as the best classifier concerning separation of both lymphoma types. Overexpression in these malignancies suggests ETV5 as a new candidate for a pathogenic factor in B-cell lymphomas. Characteristic deregulation of multiple genes analyzed in this study could be combined in a comprehensive picture of 2 distinctive pathomechanisms in B-CLL and MCL. In B-CLL, the expression parameters are in strong favor of protection of the malignant cells from apoptosis but did not provide evidence for promoting cell cycle. In contrast, in MCL the impairment of apoptosis induction seems to play a minor role, whereas most expression data indicate an enhancement of cell proliferation.