Merck
  • A comparison of sugar indicators enables a universal high-throughput sugar-1-phosphate nucleotidyltransferase assay.
A comparison of sugar indicators enables a universal high-throughput sugar-1-phosphate nucleotidyltransferase assay.
Analytical biochemistry (2008-04-05)
Rocco Moretti, Jon S Thorson
ZUSAMMENFASSUNG
A systematic comparison of six sugar indicators for their sensitivity, specificity, cross-reactivity, and suitability in the context of crude lysates revealed para-hydroxybenzoic acid hydrazide (pHBH) to be best suited for application in a plate-based phosphatase-assisted universal sugar-1-phosphate nucleotidyltransferase assay. The addition of a general phosphatase to nucleotidyltransferase reaction aliquots enabled the conversion of remaining sugar-1-phosphate to free sugar, the concentration of which could be rapidly assessed via the pHBH assay. The assay was validated using the model glucose-1-phosphate thymidylyltransferase from Salmonella enterica (RmlA) and compared favorably with a previously reported HPLC assay. This coupled discontinuous assay is quantitative, high throughput, and robust; relies only on commercially available enzymes and reagents; does not require chromatography, specialized detectors (e.g., mass or evaporative light scattering detectors), or radioisotopes; and is capable of detecting less than 5 nmol of sugar-1-phosphate. It is anticipated that this high-throughput assay system will greatly facilitate nucleotidyltransferase mechanistic and directed evolution/engineering studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Hydroxybenzhydrazid, ≥97%
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein