Merck
  • Identification of proteins interacting with lactate dehydrogenase in claw muscle of the porcelain crab <I>Petrolisthes cinctipes</I>.
Identification of proteins interacting with lactate dehydrogenase in claw muscle of the porcelain crab &lt;I&gt;Petrolisthes cinctipes&lt;/I&gt;.
Comparative Biochemistry and Physiology (2011)
Cayenne, A.P., et al.