Merck
  • The source, composition and flux of polycyclic aromatic hydrocarbons in sediments of the river Derwent, Derbyshire, U.K.
The source, composition and flux of polycyclic aromatic hydrocarbons in sediments of the river Derwent, Derbyshire, U.K.
Water, Air, Soil Pollut. (1990)
Evans, K.M., et al.