Merck
  • Unravelling evolutionary strategies of yeast for improving galactose utilization through integrated systems level analysis
Unravelling evolutionary strategies of yeast for improving galactose utilization through integrated systems level analysis
Proceedings of the National Academy of Sciences of the USA (2011)
Hong KK, et al.