Merck
  • Glutathione monoethyl ester: high-performance liquid chromatographic analysis and direct preparation of the free base form.
Glutathione monoethyl ester: high-performance liquid chromatographic analysis and direct preparation of the free base form.
Analytical biochemistry (1989-11-15)
E B Campbell, O W Griffith
ZUSAMMENFASSUNG
Glutathione monoethyl ester (L-gamma-glutamyl-L-cysteinylglycine ethyl ester) was shown by R. N. Puri and A. Meister (1983, Proc. Natl. Acad. Sci. USA 80, 5258-5260) to be taken up by several tissues and intracellularly hydrolyzed to GSH. Since GSH itself is not significantly taken up by tissues, glutathione monoesters provide the most direct and convenient means available for increasing the intracellular GSH concentration of many tissues and cell types. In previous studies glutathione esters were prepared by HCl- or H2SO4-catalyzed esterification, and the product esters were precipitated as acidic salts by addition of ether to the reaction mixtures. In the present studies, glutathione monoethyl ester was synthesized by H2SO4-catalyzed esterification in the presence of sodium sulfate as the dehydrating agent. When no GSH remained, alcohol-washed Dowex-1 resin (hydroxide form) was added to remove sulfate and neutralize the reaction mixture. After the resin was removed by filtration, glutathione monoethyl ester crystallized in the chilled filtrate. The product was free of sulfate, GSH, and glutathione diester; its solutions in water or saline were neutral. Preparations obtained to date are nontoxic when administered to mice in doses up to at least 10 mmol/kg. Progress of the esterification reaction and purity of the product were determined quantitatively by HPLC after derivatization of the thiols with monobromobimane. Elution times of GSH, glutathione diester, and glutathione monoesters involving either the glutamyl or the glycyl carboxylate groups are reported.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glutathione reduced ethyl ester, ≥90% (TLC)