Merck
  • Dynamic strength of the interaction between lung surfactant protein D (SP-D) and saccharide ligands.
Dynamic strength of the interaction between lung surfactant protein D (SP-D) and saccharide ligands.
Biochemistry (2007-10-06)
Esben Thormann, Jakob K Dreyer, Adam C Simonsen, Per L Hansen, Søren Hansen, Uffe Holmskov, Ole G Mouritsen
ZUSAMMENFASSUNG
In order to investigate the dynamic strength of the interaction between lung surfactant protein D (SP-D) and different sugars, maltose, mannose, glucose, and galactose, we have used an atomic force microscope to monitor the interaction on a single molecule scale. The experiment is performed by measuring the rupture force when the SP-D-sugar bond is subjected to a continuously increasing force. Under these dynamic conditions, SP-D binds strongest to d-mannose and weakest to maltose and d-galactose. These results differ from equilibrium measurements wherein SP-D exhibits preference for maltose. On the basis of this finding, we propose that the binding of the disaccharide maltose to SP-D, which is energetically stronger than the binding of any of the monosacchrides, alters the structure of the binding site in a way that lowers the dynamic strength of the bond. We conclude that determining the strength of a protein-ligand bond under dynamic stress using an atomic force microscope is possibly more relevant for mimicking the actual nonequilibrium physiological situation in the lungs.