Merck
  • Preparation of ready-to-use, storable and reconstituted type I collagen from rat tail tendon for tissue engineering applications
Preparation of ready-to-use, storable and reconstituted type I collagen from rat tail tendon for tissue engineering applications
Nature Protocols (2006)
Rajan N, et al.