Merck
  • Effect of enzyme treatment and mechanical removal of alpaca (Vicugna pacos) seminal plasma on sperm functional integrity
Effect of enzyme treatment and mechanical removal of alpaca (Vicugna pacos) seminal plasma on sperm functional integrity
JOURNAL-SHANDONG UNIVERSITY OF SCIENCE AND TECHNOLOGY NATURAL SCIENCE (2012)
Morton K M, et al.