Merck
  • MrkD (1P) from Klebsiella pneumoniae IA565 allows for co-existence with Pseudomonas aeruginosa and protection from protease-mediated biofilm detachment
MrkD (1P) from Klebsiella pneumoniae IA565 allows for co-existence with Pseudomonas aeruginosa and protection from protease-mediated biofilm detachment
Infection and Immunity (2013)
Childers B M, et al.