Merck
  • N-Guanidino Derivatives of 1, 5-Dideoxy-1, 5-imino-d-xylitol are Potent, Selective, and Stable Inhibitors of beta-Glucocerebrosidase
N-Guanidino Derivatives of 1, 5-Dideoxy-1, 5-imino-d-xylitol are Potent, Selective, and Stable Inhibitors of beta-Glucocerebrosidase
ChemMedChem (2017)
Sevvsek A, et al.