Merck
  • Clostridia: the importance of their exceptional substrate and metabolite diversity for biofuel and biorefinery applications.
Clostridia: the importance of their exceptional substrate and metabolite diversity for biofuel and biorefinery applications.
Current Opinion in Biotechnology (2011)
Tracy, B.P., et al.