Merck
  • DHEA improves the antioxidant capacity of endometrial stromal cells and improves endometrium receptivity via androgen receptor
DHEA improves the antioxidant capacity of endometrial stromal cells and improves endometrium receptivity via androgen receptor
Eur. J. Obstet. Gyn. Rep. Biol. (2016)
Qin A, et al.