Merck
Synthesis of full-length cDNA using DNA-capped mRNA.
Nucleic acids symposium series (1993-01-01)
S Sekine, S Kato
ZUSAMMENFASSUNG
We have developed a novel method for synthesizing a full-length cDNA based on the addition of DNA-RNA chimeric linker to the CAP site of mRNA. The DNA-capped mRNA was used as a template for cDNA synthesis with a dT-tailed vector primer. The model experiment using rabbit globin mRNA showed that more than 80% of the clones encoding alpha- and beta-globin contains a full-length cDNAs starting from the CAP site.