Merck
  • Isolation of Sertoli cells from adult rat testes: an approach to ex vivo studies of Sertoli cell function
Isolation of Sertoli cells from adult rat testes: an approach to ex vivo studies of Sertoli cell function
Biology of Reproduction (2003)
Anway MD, et al.