Merck
  • Selectivity in the potentiation of antibacterial activity of alpha-peptide beta-peptoid peptidomimetics and antimicrobial peptides by human blood plasma
Selectivity in the potentiation of antibacterial activity of alpha-peptide beta-peptoid peptidomimetics and antimicrobial peptides by human blood plasma
Research in Microbiology (2013)
Hein-Kristensen L, et al.