Merck
  • Positive reinforcing effects of substance P in the rat globus pallidus revealed by conditioned place preference
Positive reinforcing effects of substance P in the rat globus pallidus revealed by conditioned place preference
Behavioural Brain Research (2010)
Kertes, E, et al.