Merck
  • Insect-like organization of the stomatopod central complex: functional and phylogenetic implications
Insect-like organization of the stomatopod central complex: functional and phylogenetic implications
Frontiers in Behavioral Neuroscience (2017)
Thoen HH, et al.