- Oxidative degradation of the monolayer of 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine (POPC) in low-level ozone
Oxidative degradation of the monolayer of 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine (POPC) in low-level ozone
The Journal of Physical Chemistry B (2015)
Qiao L, et al.