Merck
  • Modified apolipoprotein (apo) AI by artificial sweetener causes severe premature cellular senescence and atherosclerosis with impairment of functional and structural properties of apoA-I in lipid-free and lipid-bound state
Modified apolipoprotein (apo) AI by artificial sweetener causes severe premature cellular senescence and atherosclerosis with impairment of functional and structural properties of apoA-I in lipid-free and lipid-bound state
Molecules and Cells (2011)
Jang W, et al.