- Arginase inhibition restores arteriolar endothelial function in Dahl rats with salt-induced hypertension
Arginase inhibition restores arteriolar endothelial function in Dahl rats with salt-induced hypertension
American Journal of Physiology. Lung Cellular and Molecular Physiology (2005)
Johnson FK, et al.