Merck
  • Trehalose is a key determinant of the quiescent metabolic state that fuels cell cycle progression upon return to growth
Trehalose is a key determinant of the quiescent metabolic state that fuels cell cycle progression upon return to growth
Molecular Biology of the Cell (2010)
Shi Lei, et al.