Merck
  • Stem girdling manipulates leaf sugar concentrations and anthocyanin expression in sugar maple trees during autumn
Stem girdling manipulates leaf sugar concentrations and anthocyanin expression in sugar maple trees during autumn
Tree physiology (2008)
Murakami PF, et al.