Merck
  • Integrin alphavbetabgr3-RGDS interaction mediates fibrin-induced morphological changes of glomerular endothelial cells
Integrin alphavbetabgr3-RGDS interaction mediates fibrin-induced morphological changes of glomerular endothelial cells
Kidney International (1999)
Xu Q, et al.