Merck
  • Prolonged exposure to neutrophil extracellular traps can induce mitochondrial damage in macrophages and dendritic cells
Prolonged exposure to neutrophil extracellular traps can induce mitochondrial damage in macrophages and dendritic cells
SpringerPlus (2015)
Donis-Maturano L, et al.