Merck
  • Dasatinib synergizes with doxorubicin to block growth, migration, and invasion of breast cancer cells
Dasatinib synergizes with doxorubicin to block growth, migration, and invasion of breast cancer cells
British Journal of Cancer (2009)
Pichot CS, et al.