Merck
  • Cell damage-induced conformational changes of the pro-apoptotic protein Bak in vivo precede the onset of apoptosis.
Cell damage-induced conformational changes of the pro-apoptotic protein Bak <I>in vivo</I> precede the onset of apoptosis.
Cell (1999)
Griffiths, GJ.