Merck
  • Gramicidin S: A Potent Inhibitor of Membrane-bound Epidennal Adenosine Triphosphatase from Nicotiana tabacum L. Leaves.
Gramicidin S: A Potent Inhibitor of Membrane-bound Epidennal Adenosine Triphosphatase from <I>Nicotiana tabacum</I> L. Leaves.
Plant &amp; Cell Physiology (1982)
Kunihiro Kasamo