Merck
  • Iduronic acid-containing glycosaminoglycans on target cells are required for efficient respiratory syncytial virus infection
Iduronic acid-containing glycosaminoglycans on target cells are required for efficient respiratory syncytial virus infection
Virology (2000)
Hallak LK, et al.