Merck
  • Diazeniumdiolation of protamine sulfate reverses mitogenic effects on smooth muscle cells and fibroblasts
Diazeniumdiolation of protamine sulfate reverses mitogenic effects on smooth muscle cells and fibroblasts
Free radical biology & medicine (2015)
Van Lith R, et al.