Merck
Ribosomal protection proteins and their mechanism of tetracycline resistance.
Antimicrobial agents and chemotherapy (2003-11-26)
Sean R Connell, Dobryan M Tracz, Knud H Nierhaus, Diane E Taylor
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)