Merck
  • Detection of superoxide anion radical in a stratum corneum intercellular lipid model using an electrochemical sensor
Detection of superoxide anion radical in a stratum corneum intercellular lipid model using an electrochemical sensor
Journal of Oleo Science (2011)
Kaneko T, et al.