- Rapid screening of purification strategies for the capture of a human recombinant F(ab')2 expressed in baculovirus-infected cells using a micro-plate approach and SELDI-MS.
Rapid screening of purification strategies for the capture of a human recombinant F(ab')2 expressed in baculovirus-infected cells using a micro-plate approach and SELDI-MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2009-05-27)
J Pezzini, V Brenac Brochier, M P Barrouillet, M Cerruti, G Clofent-Sanchez, A Schapman, A Topol, R Robert, C Cabanne, P Cerruti, X Santarelli
PMID19467934
ZUSAMMENFASSUNG
The development of a capture step of a human recombinant F(ab')(2) produced and expressed in baculovirus-infected cells was investigated by screening three mixed-mode chromatography sorbents (HEA HyperCel, PPA HyperCel and MEP HyperCel) and two ion exchangers (Q Ceramic HyperD F, S Ceramic HyperD F) sorbents using a 96-well plate format and SELDI-MS. HEA HyperCel gave the best separation performance therefore the conditions tested in micro-plate were transferred to laboratory scale chromatographic experiments, confirming that the recombinant F(ab')(2) was effectively captured on the mixed-mode sorbent without any pre-treatment of the crude extract with a 82% recovery and a 39-fold purification.