Merck
Plasma biomarkers in a mouse model of preterm labor.
Pediatric research (2009-03-17)
Qing Yang, John C Whitin, Xuefeng Bruce Ling, Nihar R Nayak, Harvey J Cohen, Joseph Jin, James Schilling, Tom To-Sang Yu, Ashima Madan
ZUSAMMENFASSUNG
Preterm labor (PTL) is frequently associated with inflammation. We hypothesized that biomarkers during pregnancy can identify pregnancies most at risk for development of PTL. An inflammation-induced mouse model of PTL was used. Surface-enhanced laser desorption/ionization time-of-flight mass spectrometry was used to analyze and compare the plasma protein (PP) profile between CD-1 mice injected intrauterine with either lipopolysaccharide (LPS) or PBS on d 14.5 of gestation. The median differences of normalized PP peaks between the two groups were determined using the Mann-Whitney U test and the false discovery rate. In a second series of experiments, both groups of mice were injected with a lower dose of LPS. A total of 1665 peaks were detected. Thirty peaks were highly differentially expressed (p < 0.0001) between the groups. Two 11 kDa protein peaks were identified by MALDI-TOF/TOF-MS and confirmed to be mouse serum amyloid A (SAA) 1 and 2. Plasma SAA2 levels were increased in LPS-treated animals compared with controls and in LPS-treated animals that delivered preterm vs. those that delivered at term. SAA2 has the potential to be a plasma biomarker that can identify pregnancies at risk for development of PTL.