Merck
  • Cultivation and characterization of planarian neuronal cells isolated by fluorescence activated cell sorting (FACS)
Cultivation and characterization of planarian neuronal cells isolated by fluorescence activated cell sorting (FACS)
Zoological Science (2002)
Asami M, et al.