Merck
  • Detection of hen?s egg white lysozyme in food: Comparison between a sensitive HPLC and a commercial ELISA method.
Detection of hen?s egg white lysozyme in food: Comparison between a sensitive HPLC and a commercial ELISA method.
Food Chemistry (2010)
Kerkaert B, et al.