Merck
  • HSP90 modulates actin dynamics: inhibition of HSP90 leads to decreased cell motility and impairs invasion
HSP90 modulates actin dynamics: inhibition of HSP90 leads to decreased cell motility and impairs invasion
Biochimica et Biophysica Acta - Molecular Cell Research (2011)
Taiyab A and Rao CM