Merck
  • Hydrolysis and Decomposition of Cellulose in Bro?nsted Acidic Ionic Liquids Under Mild Conditions.
Hydrolysis and Decomposition of Cellulose in Bro?nsted Acidic Ionic Liquids Under Mild Conditions.
Industrial & Engineering Chemistry Research (2009)
Amarasekara, A.S., and Owereh, O.S.