Merck
The ubiquitin-specific protease USP10 modulates androgen receptor function.
Molecular and cellular endocrinology (2005-12-22)
Hortensia Faus, Hellmuth-Alexander Meyer, Martina Huber, Inke Bahr, Bernard Haendler
ZUSAMMENFASSUNG
The role of the ubiquitin/proteasome system in degrading nuclear hormone receptors and regulating their transcriptional function has emerged in the last few years. We identified the ubiquitin-specific protease USP10 as part of DNA-bound androgen receptor (AR) complexes purified from nuclear extracts of PC-3 cells stably expressing the AR. The interaction between USP10 and the AR was confirmed by GST pull-down assays. Fluorescence microscopy documented that USP10 was localised in the nucleus and the cytoplasm. Cell-based transactivation assays in PC-3/AR cells revealed that overexpression of wild-type USP10, but not of an enzymatically inactive form, stimulated AR activity mediated by reporter constructs harbouring selective androgen response elements (AREs), non-selective steroid response elements (SREs) or the mouse mammary tumour virus (MMTV) promoter. Conversely, USP10 expression knock-down by siRNAs impaired the MMTV response to androgen. In summary, the data indicate that USP10 is a new cofactor that binds to the AR and stimulates the androgen response of target promoters. This finding underlines the role of the ubiquitin/proteasome system in modulating the AR function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ubiquitin-Specific Protease 10 human, recombinant, expressed in Sf9 cells, ≥56% (SDS-PAGE)