Merck
  • Purification and Characterization of Antioxidative Peptides from Protein Hydrolysate of Lecithin-Free Egg Yolk.
Purification and Characterization of Antioxidative Peptides from Protein Hydrolysate of Lecithin-Free Egg Yolk.
Journal of the American Oil Chemists' Society (2001)
Park, P.-J., et al.