- BNIP3 protects HepG2 cells against etoposide-induced cell death under hypoxia by an autophagy-independent pathway
BNIP3 protects HepG2 cells against etoposide-induced cell death under hypoxia by an autophagy-independent pathway
Biochemical Pharmacology (2010)
Cosse JP, et al.