Merck
  • Improved LC methods for the determination of diosmin and/or hesperidin in plant extracts and pharmaceutical formulations.
Improved LC methods for the determination of diosmin and/or hesperidin in plant extracts and pharmaceutical formulations.
Journal of Pharmaceutical and Biomedical Analysis (2001)
El-Shafae AM, et al.