Merck
  • The influence of fine excipient particles on the performance of carrier-based dry powder inhalation formulations.
The influence of fine excipient particles on the performance of carrier-based dry powder inhalation formulations.
Pharmaceutical Research (2006)
Jones MD and Price R