Merck
  • An investigation into the dispersion mechanisms of ternary dry powder inhaler formulations by the quantification of interparticulate forces.
An investigation into the dispersion mechanisms of ternary dry powder inhaler formulations by the quantification of interparticulate forces.
Pharmaceutical Research (2008)
Jones MD, et al.