Merck
  • Design, synthesis, and structure-activity relationship studies of himbacine derived muscarinic receptor antagonists.
Design, synthesis, and structure-activity relationship studies of himbacine derived muscarinic receptor antagonists.
Bioorganic & medicinal chemistry letters (1999-04-17)
D Doller, S Chackalamannil, M Czarniecki, R McQuade, V Ruperto
ZUSAMMENFASSUNG
A parallel synthesis of racemic himbacine analogs was carried out by N-alkylation of various commercially available cyclic amine derivatives with the alkylating agent 4 which bears the tricyclic unit of himbacine. Several of these analogs have potency comparable to that of himbacine, albeit lacking the desired selectivity. Structure-activity relationship studies support the existence of a hydrophobic pocket in the receptor where the piperidine ring of dihydrohimbacine binds.