Merck
Mast cell proteases.
Methods in enzymology (1981-01-01)
R G Woodbury, M T Everitt, H Neurath
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chymase human, recombinant, expressed in Pichia pastoris
Sigma-Aldrich
Hippuryl-Chlor-L-Phenylalanin