Merck
Structural elucidation and bioactivities of a novel arabinogalactan from Coreopsis tinctoria.
Carbohydrate polymers (2019-06-04)
Hang Zhang, Jing Zhong, Qian Zhang, Degang Qing, Chunyan Yan
ZUSAMMENFASSUNG
Coreopsis tinctoria is being widely cultivated in Xinjiang of China, whose consumption is known to prevent diabetes and neurodegenerative diseases. To elucidate the bioactive ingredients responsible for these benefits, the alkaline soluble crude polysaccharide (CTB) was isolated from C. tinctoria. In vitro experiments showed that the inhibition of α-amylase and α-glucosidase by CTB was 13407-fold and 906-fold higher than that by positive control, respectively. Then, a novel arabinogalactan, CTBP-1, was isolated and purified from CTB. Structural analysis showed that CTBP-1 possessed a 1,6-linked β-d-Galp and 1,5-linked α-l-Araf backbone with branches substituted at the C-3 position of the 1,6-linked β-d-Galp, and the side chains included 1,5-linked α-l-Araf, T-linked β-d-Galp and T-linked α-l-Araf. The inhibitory effects of CTBP-1 on α-amylase and α-glucosidase were 2.7 and 17.9 times that of acarbose, respectively. CTBP-1 could avoid indigestion and similar side effects. In addition, CTBP-1 remarkably inhibited the release of nitric oxide (NO), tumor necrosis factor-α (TNF-α) and interleukin-6 (IL-6) in lipopolysaccharide (LPS)-stimulated BV2 microglial cells. In summary, CTBP-1 is a novel arabinogalactan with great potential as a treatment for type 2 diabetes and Alzheimer's disease.