Merck
  • Piperine-mediated suppression of voltage-dependent Ca2+ influx and glutamate release in rat hippocampal nerve terminals involves 5HT1A receptors and G protein βγ activation.
Piperine-mediated suppression of voltage-dependent Ca2+ influx and glutamate release in rat hippocampal nerve terminals involves 5HT1A receptors and G protein βγ activation.
Food & function (2019-04-30)
Ting Yang Hsieh, Yi Chang, Su Jane Wang
ZUSAMMENFASSUNG
Piperine is the crucial alkaloid component of black pepper (Piper nigrum Linn.) and has neuroprotective effects. Because inhibition of glutamatergic excitatory neurotransmission is a possible mechanism involved in neuroprotection, we investigated the effect of piperine on the 4-aminopyridine (4-AP)-evoked release of glutamate from rat hippocampal synaptosomes. Piperine inhibited 4-AP-evoked glutamate release, and the inhibition was prevented by the chelation of extracellular Ca2+ ions and a vesicular transporter inhibitor. Piperine reduced the 4-AP-evoked elevation of intrasynaptosomal Ca2+ levels but did not affect the synaptosomal membrane potential. In the presence of ω-conotoxin MVIIC, an N- and P/Q-type channel blocker, the piperine-mediated inhibition of 4-AP-evoked glutamate release was markedly reduced; however, dantrolene and CGP37157, which are intracellular Ca2+-release inhibitors, did not alter the piperine effect. In addition, immunocytochemical analysis confirmed the presence of presynaptic 5-hydroxytryptamine 1A (5-HT1A) receptor proteins. The glutamate release-inhibiting effect of piperine was discovered to be prevented by the 5-HT1A receptor antagonist WAY100635 and the G protein βγ subunit inhibitor gallein; however, it was unaffected by the adenylate cyclase inhibitor SQ22536 or the protein kinase A inhibitor PKI622. These results suggest that piperine inhibits glutamate release from rat hippocampal nerve terminals by reducing Ca2+ influx through N- and P/Q-type Ca2+ channels and that the activation of presynaptic 5-HT1A receptors and the G protein βγ subunit is involved in this effect.