Merck
Generation of a zebrafish SWATH-MS spectral library to quantify 10,000 proteins.
Scientific data (2019-02-13)
Peter Blattmann, Vivienne Stutz, Giulia Lizzo, Joy Richard, Philipp Gut, Ruedi Aebersold
ZUSAMMENFASSUNG
Sequential window acquisition of all theoretical mass spectra (SWATH-MS) requires a spectral library to extract quantitative measurements from the mass spectrometry data acquired in data-independent acquisition mode (DIA). Large combined spectral libraries containing SWATH assays have been generated for humans and several other organisms, but so far no publicly available library exists for measuring the proteome of zebrafish, a rapidly emerging model system in biomedical research. Here, we present a large zebrafish SWATH spectral library to measure the abundance of 104,185 proteotypic peptides from 10,405 proteins. The library includes proteins expressed in 9 different zebrafish tissues (brain, eye, heart, intestine, liver, muscle, ovary, spleen, and testis) and provides an important new resource to quantify 40% of the protein-coding zebrafish genes. We employ this resource to quantify the proteome across brain, muscle, and liver and characterize divergent expression levels of paralogous proteins in different tissues. Data are available via ProteomeXchange (PXD010876, PXD010869) and SWATHAtlas (PASS01237).