Merck
Structural and functional consequences of the STAT5BN642H driver mutation.
Nature communications (2019-06-09)
Elvin D de Araujo, Fettah Erdogan, Heidi A Neubauer, Deniz Meneksedag-Erol, Pimyupa Manaswiyoungkul, Mohammad S Eram, Hyuk-Soo Seo, Abdul K Qadree, Johan Israelian, Anna Orlova, Tobias Suske, Ha T T Pham, Auke Boersma, Simone Tangermann, Lukas Kenner, Thomas Rülicke, Aiping Dong, Manimekalai Ravichandran, Peter J Brown, Gerald F Audette, Sarah Rauscher, Sirano Dhe-Paganon, Richard Moriggl, Patrick T Gunning
ZUSAMMENFASSUNG
Hyper-activated STAT5B variants are high value oncology targets for pharmacologic intervention. STAT5BN642H, a frequently-occurring oncogenic driver mutation, promotes aggressive T-cell leukemia/lymphoma in patient carriers, although the molecular origins remain unclear. Herein, we emphasize the aggressive nature of STAT5BN642H in driving T-cell neoplasia upon hematopoietic expression in transgenic mice, revealing evidence of multiple T-cell subset organ infiltration. Notably, we demonstrate STAT5BN642H-driven transformation of γδ T-cells in in vivo syngeneic transplant models, comparable to STAT5BN642H patient γδ T-cell entities. Importantly, we present human STAT5B and STAT5BN642H crystal structures, which propose alternative mutation-mediated SH2 domain conformations. Our biophysical data suggests STAT5BN642H can adopt a hyper-activated and hyper-inactivated state with resistance to dephosphorylation. MD simulations support sustained interchain cross-domain interactions in STAT5BN642H, conferring kinetic stability to the mutant anti-parallel dimer. This study provides a molecular explanation for the STAT5BN642H activating potential, and insights into pre-clinical models for targeted intervention of hyper-activated STAT5B.